missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine Hydrochloride, 98%, Spectrum™ Chemical

Catalog No. 18610231
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610231 100 g Amber Glass Bottle
18610235 500 g Amber Glass Bottle
18610233 2.5 kg Amber Glass Bottle
18610232 12 kg Amber Glass Bottle
18610234 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610231 Supplier Spectrum Chemical Mfg Cor Supplier No. D2396100GMGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 506-59-2

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 g
Molecular Weight (g/mol) 81.55
Percent Purity 0.98
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Dimethylamine Hydrochloride

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.