missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine, MilliporeSigma™

Catalog No. MDX10455
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle (Pre-Packed)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MDX10455 500 mL Glass Bottle (Pre-Packed)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MDX10455 Supplier Technidata Supplier No. DX10455

Chemical Identifiers

CAS 109-89-7
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00009032
InChI Key HPNMFZURTQLUMO-UHFFFAOYSA-N
Synonym diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine
PubChem CID 8021
ChEBI CHEBI:85259
IUPAC Name diethylamine
SMILES CCNCC

Specifications

Density 0.71g/cm3 (20°C)
Explosion Limit 2.0 to 11.8%(V)
Ignition Point 310°C
Packaging Glass Bottle (Pre-Packed)
Linear Formula (C2H5)2NH
Quantity 500 mL
Vapor Pressure 250 hPa (20°C)
Formula Weight 73.14
Chemical Name or Material Diethylamine
Recommended Storage Store at +15°C to +25°C