missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine Borane 95.0+%, TCI America™

Catalog No. p-7126964
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D184225G 25 g
D1842100G 100 g
D1842500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D184225G Supplier TCI America Supplier No. D184225G
Only null left

Chemical Identifiers

CAS 74-94-2
Molecular Formula C2H10BN
Molecular Weight (g/mol) 58.92
MDL Number MFCD00051068
InChI Key FTDUHBOCJSQEKS-UHFFFAOYSA-N
Synonym Borane - Dimethylamine Complex, DMAB
IUPAC Name dimethylamine borane
SMILES B.CNC

Specifications

Melting Point 37°C
Color White
Quantity 25 g
UN Number 2926
Formula Weight 58.92
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylamine Borane
EINECSNumber (2)-2102
RTECSNumber IP9450000
TSCA Yes
Recommended Storage Refrigerator