missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylamine Borane 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 37°C |
| Color | White |
| Quantity | 500 g |
| UN Number | 2926 |
| Formula Weight | 58.92 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dimethylamine Borane |
Safety and Handling
| EINECSNumber | (2)-2102 |
| RTECSNumber | IP9450000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |