Abcam Inc Dimethylamiloride hydrochloride (5-(N,N-Dimethyl)amiloride hydrochloride), Na+/H+ e x change blocker, 100MG
Catalog No. 502705136
MW 294.14 Da, Purity >97%. Selectively inhibits Na+/H+ exchangers in cellular membranes, resulting in the retention of H+ ions and in the decrease of intracellular pH. Also blocks TRPA1 channels (IC50 = 80 M).
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 294.14 Da, Purity >97%. Selectively inhibits Na+/H+ exchangers in cellular membranes, resulting in the retention of H+ ions and in the decrease of intracellular pH. Also blocks TRPA1 channels (IC50 = 80 M).