Learn More
Dimethylaluminium chloride, 0.9M solution in heptane, AcroSeal™
CAS: 1184-58-3 | C2H6AlCl | 92.502 g/mol
$203.88 - $203.88
Chemical Identifiers
| CAS | 1184-58-3 |
|---|---|
| MDL Number | MFCD00000458 |
| InChI Key | JGHYBJVUQGTEEB-UHFFFAOYSA-M |
| Synonym | aluminum, chlorodimethyl, dimethylaluminum chloride, dimethylaluminium chloride, chlorodimethylaluminum, dimethylaluminum chloride solution, 1.0 m in hexanes, chloro dimethyl alumane, dimethylaluminum chloride solution |
| PubChem CID | 79147 |
| IUPAC Name | chloro(dimethyl)alumane |
| SMILES | C[Al](C)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC377791000
|
Thermo Scientific Chemicals
377791000 |
100 mL | AcroSeal™ Glass bottle |
Each for $203.88
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
| 1184-58-3 | |
| JGHYBJVUQGTEEB-UHFFFAOYSA-M | |
| 79147 | |
| C[Al](C)Cl |
| MFCD00000458 | |
| aluminum, chlorodimethyl, dimethylaluminum chloride, dimethylaluminium chloride, chlorodimethylaluminum, dimethylaluminum chloride solution, 1.0 m in hexanes, chloro dimethyl alumane, dimethylaluminum chloride solution | |
| chloro(dimethyl)alumane |
Specifications
| 1184-58-3,142-82-5 | |
| 0.7110g/mL | |
| AcroSeal™ Glass bottle | |
| MFCD00000458 | |
| 0.711 | |
| aluminum, chlorodimethyl, dimethylaluminum chloride, dimethylaluminium chloride, chlorodimethylaluminum, dimethylaluminum chloride solution, 1.0 m in hexanes, chloro dimethyl alumane, dimethylaluminum chloride solution | |
| JGHYBJVUQGTEEB-UHFFFAOYSA-M | |
| chloro(dimethyl)alumane | |
| 79147 | |
| Solution |
| Colorless to Gray | |
| −4°C | |
| C2H6AlCl | |
| 100 mL | |
| 15, 3249 | |
| Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons | |
| C[Al](C)Cl | |
| 92.5 | |
| 92.5 | |
| Dimethylaluminium chloride, 0.9M Solution in Heptane |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
In contact with water releases flamm
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap
GHS Signal Word: Danger
EINECSNumber : 214-668-8
RUO – Research Use Only