missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl trans-4-Cyclohexene-1,2-dicarboxylate 98.0+%, TCI America™

Catalog No. p-7127867
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36921G 1 g
D36925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36921G Supplier TCI America Supplier No. D36921G

Chemical Identifiers

CAS 17673-68-6
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
InChI Key DVVAGRMJGUQHLI-HTQZYQBOSA-N
Synonym trans-4-Cyclohexene-1,2-dicarboxylic Acid Dimethyl Ester
PubChem CID 253379
IUPAC Name dimethyl (1R,2R)-cyclohex-4-ene-1,2-dicarboxylate
SMILES COC(=O)C1CC=CCC1C(=O)OC

Specifications

Color Yellow
Boiling Point 119°C
Quantity 1 g
Formula Weight 198.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl trans-4-Cyclohexene-1,2-dicarboxylate
EINECSNumber (3)-2437
TSCA No