missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, Analytical Standard, ≥99.95% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7193451
Change view
Click to view available options
Quantity:
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020405 1 mL
111018317 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020405 Supplier Merck Emd Millipore Supplier No. 945631MLF
Only null left

For information on dimethyl sulfoxide miscibility, please visit the following link:Dimethyl Sulfoxide Miscibility/Immiscibility Table

Specifications

Melting Point 16°C to 19°C
CAS 67-68-5
Density 1.10 g/mL
Quantity 1 mL
Physical Form Neat
Linear Formula (CH3)2 SO
MDL Number MFCD00002089
Vapor Pressure 0.42 mmHg (20°C)
Synonym DMSO
Solubility Information H2O: miscible (completely)
Formula Weight 78.13
Percent Purity ≥99.95% (GC)
Show More Show Less