Learn More
Merck Emd Millipore Dimethyl sulfoxide Hybri-Max, 99.7, 67-68-5, MFCD00002089, 100mL
Supplier: Merck Emd Millipore D2650100ML
Linear Formula (CH3)2SO, 99.7%, Molecular Weight 78.13, Hybri-Max™, sterile-filtered, BioReagent, Synonym: DMSO