missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfone 99.0+%, TCI America™

Catalog No. p-7132611
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M123925G 25 g
M1239500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M123925G Supplier TCI America Supplier No. M123925G
Only null left

Chemical Identifiers

CAS 67-71-0
Molecular Formula C2H6O2S
Molecular Weight (g/mol) 94.13
MDL Number MFCD00007566
InChI Key HHVIBTZHLRERCL-UHFFFAOYSA-N
Synonym dimethyl sulfone, methyl sulfone, sulfonylbismethane, dimethylsulfone, dimethyl sulphone, methane, sulfonylbis, sulphonylbismethane, methylsulfone, methanesulfonylmethane, sulfonyldimethane
PubChem CID 6213
ChEBI CHEBI:9349
IUPAC Name methanesulfonylmethane
SMILES CS(C)(=O)=O

Specifications

Melting Point 110°C
Color White-Yellow
Boiling Point 238°C
Quantity 25 g
Formula Weight 94.13
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Sulfone
EINECSNumber (2)-1557
RTECSNumber PB2785000
TSCA Yes