missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfite 99.0+%, TCI America™

Catalog No. D026425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D026425G 25 g
D0264500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D026425G Supplier TCI America Supplier No. D026425G
Only null left

Chemical Identifiers

CAS 616-42-2
Molecular Formula C2H6O3S
Molecular Weight (g/mol) 110.127
MDL Number MFCD00008415
InChI Key BDUPRNVPXOHWIL-UHFFFAOYSA-N
Synonym methyl sulfite, sulfurous acid, dimethyl ester, dimethyl sulphite, dimethoxy sulfoxide, dimethylsulfit, unii-9jfa40s66p, ch3o 2so, sulphurous acid dimethyl ester, dimethyl ester of sulfurous acid, dimethylsulfite
PubChem CID 69223
ChEBI CHEBI:48858
IUPAC Name dimethyl sulfite
SMILES COS(=O)OC

Specifications

Color Colorless
Boiling Point 126°C
Quantity 25 g
UN Number 1993
Formula Weight 110.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Sulfite
RTECSNumber WT3512000
TSCA Yes