missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfite, 99%

CAS: 616-42-2 | C2H6O3S | 110.13 g/mol

Supplier:  Thermo Scientific Chemicals 148751000

Catalog No. AC148751000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

Dimethyl sulfite
616-42-2
1.2900g/mL
30°C
99%
C2H6O3S
(CH3O)2SO
100 mL
01,296
methyl sulfite, sulfurous acid, dimethyl ester, dimethyl sulphite, dimethoxy sulfoxide, dimethylsulfit, unii-9jfa40s66p, ch3o 2so, sulphurous acid dimethyl ester, dimethyl ester of sulfurous acid, dimethylsulfite
BDUPRNVPXOHWIL-UHFFFAOYSA-N
dimethyl sulfite
69223
110.13
Liquid
99%
Colorless to Yellow
126°C to 127°C
Authentic
Glass bottle
1.4085 to 1.4105
MFCD00008415
01, 282
1.29
Solubility in water: soluble. Other solubilities: soluble in dmf,slightly soluble in nonpolar solvents
COS(=O)OC
110.13
CHEBI:48858
99%

Chemical Identifiers

616-42-2
110.13
BDUPRNVPXOHWIL-UHFFFAOYSA-N
69223
dimethyl sulfite
C2H6O3S
MFCD00008415
methyl sulfite, sulfurous acid, dimethyl ester, dimethyl sulphite, dimethoxy sulfoxide, dimethylsulfit, unii-9jfa40s66p, ch3o 2so, sulphurous acid dimethyl ester, dimethyl ester of sulfurous acid, dimethylsulfite
CHEBI:48858
COS(=O)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-481-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only