missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl selenide

CAS: 593-79-3 | C2H6Se | 109.041 g/mol

$128.51 - $1493.45

Chemical Identifiers

CAS 593-79-3
Molecular Formula C2H6Se
Molecular Weight (g/mol) 109.041
MDL Number MFCD00014848
InChI Key RVIXKDRPFPUUOO-UHFFFAOYSA-N
Synonym dimethylselenide, dimethylselenium, dimethyl selenide, methyl selenide, methyl selenium, methane, selenobis, selenium dimethyl, selenide, dimethyl, ch3 2se, unii-yk0r6jkt6h
PubChem CID 11648
ChEBI CHEBI:4610
IUPAC Name methylselanylmethane
SMILES C[Se]C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA6811603
View Documents
Thermo Scientific Chemicals
06811603
1 g
Each for $128.51
Only null left
 
AA6811606
View Documents
Thermo Scientific Chemicals
06811606
5 g
Each for $441.33
Only null left
 
AA6811614
View Documents
Thermo Scientific Chemicals
06811614
25 g
Each for $1,493.45
Only null left
 
Description

Description

Used as primary and secondary intermediate and also used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

593-79-3
109.041
RVIXKDRPFPUUOO-UHFFFAOYSA-N
11648
methylselanylmethane
C2H6Se
MFCD00014848
dimethylselenide, dimethylselenium, dimethyl selenide, methyl selenide, methyl selenium, methane, selenobis, selenium dimethyl, selenide, dimethyl, ch3 2se, unii-yk0r6jkt6h
CHEBI:4610
C[Se]C

Specifications

593-79-3
1.41 g/mL
Unpleasant
(CH3)2Se
1 g
1696848
dimethylselenide, dimethylselenium, dimethyl selenide, methyl selenide, methyl selenium, methane, selenobis, selenium dimethyl, selenide, dimethyl, ch3 2se, unii-yk0r6jkt6h
RVIXKDRPFPUUOO-UHFFFAOYSA-N
methylselanylmethane
11648
109.03
Dimethyl selenide
Colorless
56°C to 58°C
C2H6Se
MFCD00014848
UN3282
Air and light sensitive
Very soluble in ethyl ether and ethyl alcohol.
C[Se]C
109.041
CHEBI:4610
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H301-H330-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC COMPOUND, TOXIC, LIQUID, N.O.S.

EINECSNumber : 209-807-4

RTECSNumber : VS6880000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only