missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl pyridine-2,6-dicarboxylate, 98%

Catalog No. AAH3225118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3225118 50 g
AAH3225109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3225118 Supplier Thermo Scientific Chemicals Supplier No. H3225118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5453-67-8 | C9H9NO4 | 195.174 g/mol

Eight macrocyclic bisamides and tetramides have been synthesized by reaction of dimethyl pyridine-2,6-dicarboxylate 1 with α,ω-diaminoethers in methanol as a solvent. Silver(I) coordination polymer and nine-coordinated cadmium(II) complex with dimethyl pyridine-2,6-dicarboxylate supported by solid state and electrochemical studies

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Eight macrocyclic bisamides and tetramides have been synthesized by reaction of dimethyl pyridine-2,6-dicarboxylate 1 with α,ω-diaminoethers in methanol as a solvent. Silver(I) coordination polymer and nine-coordinated cadmium(II) complex with dimethyl pyridine-2,6-dicarboxylate supported by solid state and electrochemical studies

Solubility
Soluble in water(14 g/L) (25°C). Soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 5453-67-8
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00134493
InChI Key SNQQJEJPJMXYTR-UHFFFAOYSA-N
Synonym dimethyl 2,6-pyridinedicarboxylate, 2,6-dimethyl pyridine-2,6-dicarboxylate, pyridine-2,6-dicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid, dimethyl ester, dimethyl dipicolinate, dimethyl pyridine-2,6-carboxylate, chembl72405, dimethylpyridine-2,6-dicarboxylate, dipicolinic acid dimethyl ester
PubChem CID 79549
IUPAC Name dimethyl pyridine-2,6-dicarboxylate
SMILES COC(=O)C1=NC(=CC=C1)C(=O)OC

Specifications

Melting Point 119°C to 122°C
Quantity 50 g
Solubility Information Soluble in water(14g/L) (25°C). Soluble in chloroform.
Formula Weight 195.17
Percent Purity 98%
Chemical Name or Material Dimethyl pyridine-2,6-dicarboxylate
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 226-697-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only