missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl pyridine-2,5-dicarboxylate, 97%

CAS: 881-86-7 | C9H9NO4 | 195.174 g/mol

Supplier:  Thermo Scientific Chemicals A1725014

 View more versions of this product

Catalog No. AAA1725014


Only null left
Explore available promotions

Description

Description

It is the intermediate in the synthesis of potent antitumor agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the intermediate in the synthesis of potent antitumor agents.

Solubility
Soluble in Dichloromethane and Ethyl Acetate. Insoluble in water.

Notes
Store at -20°C. Incompatible with oxidizing agents, reducing agents, bases.
Specifications

Specifications

Dimethyl pyridine-2,5-dicarboxylate
160°C to 163°C
MFCD00034767
172251
Soluble in Dichloromethane and Ethyl Acetate. Insoluble in water.
COC(=O)C1=CN=C(C=C1)C(=O)OC
195.174
195.17
881-86-7
C9H9NO4
25 g
dimethyl 2,5-pyridinedicarboxylate, 2,5-pyridinedicarboxylic acid, dimethyl ester, dimethylpyridine-2,5-dicarboxylate, 2,5-dimethyl pyridine-2,5-dicarboxylate, dimethyl isocinchomeronate, pyridine-2,5-dicarboxylic acid dimethyl ester, dimethyl 2,5-pyridine dicarboxylate, acmc-1bkgx, maybridge1_006391, methyl 6-methoxycarbonylnicotinate
TUGSJNQAIMFEDY-UHFFFAOYSA-N
dimethyl pyridine-2,5-dicarboxylate
234911
≥98%

Chemical Identifiers

881-86-7
195.174
TUGSJNQAIMFEDY-UHFFFAOYSA-N
234911
COC(=O)C1=CN=C(C=C1)C(=O)OC
C9H9NO4
MFCD00034767
dimethyl 2,5-pyridinedicarboxylate, 2,5-pyridinedicarboxylic acid, dimethyl ester, dimethylpyridine-2,5-dicarboxylate, 2,5-dimethyl pyridine-2,5-dicarboxylate, dimethyl isocinchomeronate, pyridine-2,5-dicarboxylic acid dimethyl ester, dimethyl 2,5-pyridine dicarboxylate, acmc-1bkgx, maybridge1_006391, methyl 6-methoxycarbonylnicotinate
dimethyl pyridine-2,5-dicarboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only