missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl pyridine-2,3-dicarboxylate, 95%

Catalog No. p-7056170
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6493906 5 g
AAH6493914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6493906 Supplier Thermo Scientific Chemicals Supplier No. H6493906

CAS: 605-38-9 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 605-38-9
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00186549
InChI Key YLGIBCYHQZTFQL-UHFFFAOYSA-N
Synonym 2,3-pyridinedicarboxylic acid dimethyl ester, dimethyl quinolinate, dimethyl 2,3-pyridinedicarboxylate, 2,3-bis methoxycarbonyl pyridine, 2,3-pyridinedicarboxylic acid, dimethyl ester, methylquinolinic acid, 2,3-dimethyl ester, 2,3-dimethyl pyridine-2,3-dicarboxylate, pyridine-2,3-dicarboxylic acid dimethyl ester, mqdme, 2,3-dimethyl methylquinolinate
PubChem CID 69058
IUPAC Name dimethyl pyridine-2,3-dicarboxylate
SMILES COC(=O)C1=CC=CN=C1C(=O)OC

Specifications

Melting Point 53°C to 58°C
Quantity 5 g
Beilstein 159846
Formula Weight 195.17
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Dimethyl pyridine-2,3-dicarboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only