missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Phenylphosphonite 98.0+%, TCI America™

Catalog No. D50141G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50141G 1 g
D50145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50141G Supplier TCI America Supplier No. D50141G

Chemical Identifiers

CAS 2946-61-4
Molecular Formula C8H11O2P
Molecular Weight (g/mol) 170.15
MDL Number MFCD00008352
InChI Key LMZLQYYLELWCCW-UHFFFAOYSA-N
Synonym dimethyl phenylphosphonite, dimethoxyphenylphosphine, phenyldimethoxyphosphine, dimethylphenylphosphonite, phosphorus, dimethoxy phenyl, phosphonous acid, phenyl-, dimethyl ester, phenylphosphonous acid dimethyl ester, acmc-20al6n, php ome 2, dimethoxy phenyl phosphane
PubChem CID 76277
IUPAC Name dimethyl phenylphosphonite
SMILES COP(OC)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 102°C
Quantity 1 g
UN Number 1760
Formula Weight 170.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Phenylphosphonite
TSCA No
Recommended Storage Refrigerator