missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Oxalate 99.0+%, TCI America™

Catalog No. p-7134176
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O008025G 25 g
O0080500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O008025G Supplier TCI America Supplier No. O008025G
Only null left

Chemical Identifiers

CAS 553-90-2
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00008442
InChI Key LOMVENUNSWAXEN-UHFFFAOYSA-N
Synonym methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid
PubChem CID 11120
ChEBI CHEBI:6859
IUPAC Name dimethyl oxalate
SMILES COC(=O)C(=O)OC

Specifications

Melting Point 54°C
Color White
Boiling Point 164°C
Quantity 25 g
Formula Weight 118.09
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Oxalate
EINECSNumber (2)-0924
RTECSNumber RO2850000
TSCA Yes