missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diméthyl Oxalate 99,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 54°C |
| Color | Blanc |
| Boiling Point | 164°C |
| Quantity | 25 g |
| Formula Weight | 118.09 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | Dimethyl Oxalate |
Sécurité et manipulation
| EINECSNumber | (2)-0924 |
| RTECSNumber | RO2850000 |
| TSCA | Yes |