missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl oxalate, 99%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Density | 1.1400g/mL |
| Boiling Point | 163°C |
| Flash Point | 75°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Plastic bottle |
| Linear Formula | CH3O2CCO2CH3 |
| Quantity | 100g |
| Beilstein | 02, 534 |
| Specific Gravity | 1.14 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. Wear protective gloves/protective clothing/eye protec |
| EINECSNumber | 209-053-6 |
| RTECSNumber | RO2850000 |
| TSCA | TSCA |
RUO – Research Use Only