Learn More
Dimethyl oxalate, 99%
CAS: 553-90-2 | C4H6O4 | 118.09 g/mol
Supplier: Thermo Scientific Chemicals A153590E
Description
Dimethyl oxalate is used as an alternate fuel in the direct oxidation fuel cell. It is involved in the catalytic hydrogenation using Cu/SiO2 to prepare ethylene glycol. It is also used in the selective gas-phase hydrogenation catalyzed by Ag/SiO2 to get the corresponding alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethyl oxalate is used as an alternate fuel in the direct oxidation fuel cell. It is involved in the catalytic hydrogenation using Cu/SiO2 to prepare ethylene glycol. It is also used in the selective gas-phase hydrogenation catalyzed by Ag/SiO2 to get the corresponding alcohol.
Solubility
Soluble in ethanol. Slightly soluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids, bases and reducing agents.
Specifications
| Dimethyl oxalate | |
| 50°C to 54°C | |
| 162°C to 163°C | |
| C4H6O4 | |
| 2500 g | |
| 14,6106 | |
| Soluble in ethanol. Slightly soluble in water. | |
| COC(=O)C(=O)OC | |
| 118.09 | |
| CHEBI:6859 | |
| 99% |
| 553-90-2 | |
| 1.148 | |
| 75°C (167°F) | |
| MFCD00008442 | |
| 1071744 | |
| methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid | |
| LOMVENUNSWAXEN-UHFFFAOYSA-N | |
| dimethyl oxalate | |
| 11120 | |
| 118.09 |
Chemical Identifiers
| 553-90-2 | |
| 118.09 | |
| LOMVENUNSWAXEN-UHFFFAOYSA-N | |
| 11120 | |
| dimethyl oxalate |
| C4H6O4 | |
| MFCD00008442 | |
| methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid | |
| CHEBI:6859 | |
| COC(=O)C(=O)OC |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 209-053-6
RTECSNumber : RO2850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only