missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Octafluoroadipate 97.0+%, TCI America™

Catalog No. p-7127809
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35901G 1 g
D35905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35901G Supplier TCI America Supplier No. D35901G
Only null left

Chemical Identifiers

CAS 3107-98-0
Molecular Formula C8H6F8O4
Molecular Weight (g/mol) 318.119
MDL Number MFCD00042267
InChI Key XPXVIIILXUOEQA-UHFFFAOYSA-N
Synonym dimethyl octafluoroadipate, dimethyl perfluoroadipate, hexanedioic acid, octafluoro-, dimethyl ester, 1,6-dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate, dimethyloctafluoroadipate, acmc-1cson, dimethyl octafluoroadipinate, dimethyl octafluorohexanedioate, perfluoroadipic acid dimethyl ester, octafluoroadipic acid dimethyl ester
PubChem CID 2737117
IUPAC Name dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate
SMILES COC(=O)C(C(C(C(C(=O)OC)(F)F)(F)F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 123°C
Quantity 1 g
Formula Weight 318.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Octafluoroadipate