missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl N-(tert-Butoxycarbonyl)-L-glutamate 98.0+%, TCI America™

Catalog No. p-7128328
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45361G 1 g
D45365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45361G Supplier TCI America Supplier No. D45361G

Chemical Identifiers

CAS 59279-60-6
Molecular Formula C12H21NO6
Molecular Weight (g/mol) 275.301
MDL Number MFCD03094774
InChI Key QNSPKWUAZQIIGZ-QMMMGPOBSA-N
Synonym boc-glu ome-ome, r-n-boc-glutamic acid-1,5-dimethyl ester, s-dimethyl 2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid dimethyl ester, s-2-tert-butoxycarbonylaminopentanedioic acid dimethyl ester, dimethyl n-tert-butoxycarbonyl-l-glutamate, dimethyl 2s-2-2-methylpropan-2-yl oxycarbonylamino pentanedioate, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, dimethyl ester, 1,5-dimethyl 2s-2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid 1,5-dimethyl ester
PubChem CID 2734644
IUPAC Name dimethyl (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioate
SMILES CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)OC

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 1 g
Formula Weight 275.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl N-(tert-Butoxycarbonyl)-L-glutamate