missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl-n-octylchlorosilane 96.0+%, TCI America™

Catalog No. D182725ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D182725ML 25 mL
D1827100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D182725ML Supplier TCI America Supplier No. D182725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18162-84-0
Molecular Formula C10H23ClSi
Molecular Weight (g/mol) 206.829
MDL Number MFCD00000511
InChI Key DBKNGKYVNBJWHL-UHFFFAOYSA-N
Synonym n-octyldimethylchlorosilane, chlorodimethyloctylsilane, octyldimethylchlorosilane, chlorodimethyl octyl silane, silane, chlorodimethyloctyl, chloro dimethyl octylsilane, dimethyloctylchlorosilane, dimethyl-n-octylchlorosilane, dimethyloctylsilyl chloride, acmc-1bvuc
PubChem CID 87482
IUPAC Name chloro-dimethyl-octylsilane
SMILES CCCCCCCC[Si](C)(C)Cl

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 mL
UN Number 2987
Formula Weight 206.83
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl-n-octylchlorosilane
EINECSNumber (2)-2041
TSCA Yes