missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl(n-octyl)(3-sulfopropyl)ammonium Hydroxide Inner Salt 98.0+%, TCI America™

Catalog No. d42475g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D42475G Supplier TCI America Supplier No. D42475G

Chemical Identifiers

CAS 15178-76-4
Molecular Formula C13H29NO3S
Molecular Weight (g/mol) 279.439
MDL Number MFCD00036912
InChI Key QZRAABPTWGFNIU-UHFFFAOYSA-N
Synonym octyl sulfobetaine, 3-dimethyl-octylazaniumyl propane-1-sulfonate, n-octyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-n,n-dimethyloctylammonio propanesulfonate inner salt, 3-dimethyl octyl ammonio propane-1-sulfonate, 3-n,n-dimethyloctylammonio propanesulfonate solution, n-octyl-n,n-dimethyl-3-ammonio-1-propanesulfonate sb-8, n-octylsulfobetaine, zwittergent 3-08
PubChem CID 3084219
IUPAC Name 3-[dimethyl(octyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]

Specifications

Melting Point 205°C
Color White
Quantity 5 g
Formula Weight 279.44
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl(n-octyl)(3-sulfopropyl)ammonium Hydroxide Inner Salt [for Biochemical Research]