missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Malonate 98.0+%, TCI America™

Catalog No. M0030500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0030500G 500 g
M003025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0030500G Supplier TCI America Supplier No. M0030500G

Chemical Identifiers

CAS 108-59-8
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00008460
InChI Key BEPAFCGSDWSTEL-UHFFFAOYSA-N
Synonym dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester
PubChem CID 7943
IUPAC Name dimethyl propanedioate
SMILES COC(=O)CC(=O)OC

Specifications

Melting Point -62°C
Color Colorless
Boiling Point 184°C
Quantity 500 g
Formula Weight 132.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Malonate
EINECSNumber (2)-0913&(2)-1107
RTECSNumber OO0950000
TSCA Yes