missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Maleate 97.0+%, TCI America™

Catalog No. p-7131926
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M001225ML 25 mL
M0012500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M001225ML Supplier TCI America Supplier No. M001225ML

Chemical Identifiers

CAS 624-48-6
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00008459
InChI Key LDCRTTXIJACKKU-ARJAWSKDSA-N
Synonym dimethyl maleate, maleic acid, dimethyl ester, methyl maleate, sipomer dmm, maleic acid dimethyl ester, dimethyl cis-butenedioate, 2-butenedioic acid z-, dimethyl ester, dimethyl cis-ethylenedicarboxylate, 2-butenedioic acid 2z-, dimethyl ester, 2-butenedioic acid, dimethyl ester, z
PubChem CID 5271565
ChEBI CHEBI:35460
IUPAC Name dimethyl (Z)-but-2-enedioate
SMILES COC(=O)C=CC(=O)OC

Specifications

Melting Point -19°C
Color Yellow
Boiling Point 205°C
Quantity 25 mL
UN Number 2810
Formula Weight 144.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Maleate
EINECSNumber (2)-1107&(2)-1142
RTECSNumber EM6300000
TSCA Yes