missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl L-(+)-Tartrate 98.0+%, TCI America™

Catalog No. T000625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T000625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T000625G Supplier TCI America Supplier No. T000625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 608-68-4
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00064437
InChI Key PVRATXCXJDHJJN-QWWZWVQMSA-N
Synonym +-dimethyl l-tartrate, dimethyl l-tartrate, 2r,3r-dimethyl 2,3-dihydroxysuccinate, dimethyl +-tartrate, l-+-tartaric acid dimethyl ester, dimethyl l-+-tartrate, 1,4-dimethyl 2r,3r-2,3-dihydroxybutanedioate, butanedioic acid, 2,3-dihydroxy-2r,3r-, 1,4-dimethyl ester, dimethyl 2r,3r-2,3-dihydroxybutanedioate, l-tartaric acid, dimethyl ester
PubChem CID 11851
IUPAC Name dimethyl (2R,3R)-2,3-dihydroxybutanedioate
SMILES COC(=O)C(C(C(=O)OC)O)O

Specifications

Melting Point 62°C
Color White
Boiling Point 118°C
Quantity 25 g
Formula Weight 178.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl L-(+)-Tartrate