missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dimethyl L-tartrate, 99%, ACROS Organics™

Catalog No. p-3778430
Change view
Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172490250 25g Glass bottle
AC172491000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC172490250 Supplier Thermo Scientific Chemicals Supplier No. 172490250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 608-68-4
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00064437
InChI Key PVRATXCXJDHJJN-QWWZWVQMSA-N
Synonym +-dimethyl l-tartrate, dimethyl l-tartrate, 2r,3r-dimethyl 2,3-dihydroxysuccinate, dimethyl +-tartrate, l-+-tartaric acid dimethyl ester, dimethyl l-+-tartrate, 1,4-dimethyl 2r,3r-2,3-dihydroxybutanedioate, butanedioic acid, 2,3-dihydroxy-2r,3r-, 1,4-dimethyl ester, dimethyl 2r,3r-2,3-dihydroxybutanedioate, l-tartaric acid, dimethyl ester
PubChem CID 11851
IUPAC Name dimethyl (2R,3R)-2,3-dihydroxybutanedioate
SMILES COC(=O)C(C(C(=O)OC)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Boiling Point 157°C to 159°C (15.0 hPa)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98+% min. (GC)
Packaging Glass bottle
Linear Formula [-CH(OH)CO2CH3]2
Quantity 25g
Beilstein 03, 510
Fieser 13,124
Specific Rotation Condition + 21.00 (20.00°C c=10,H2O)
Specific Rotation + 21
Solubility Information Solubility in water: soluble in water
Formula Weight 178.14
Percent Purity 98%
Chemical Name or Material (+)-Dimethyl L-tartrate
Show More Show Less
EINECSNumber 210-166-8

RUO – Research Use Only