missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl L-(-)-Malate 98.0+%, TCI America™

Catalog No. M13435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13435G 5 g
M134325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13435G Supplier TCI America Supplier No. M13435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 617-55-0
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00066215
InChI Key YSEKNCXYRGKTBJ-UHFFFAOYNA-N
Synonym s-dimethyl 2-hydroxysuccinate, dimethyl s---malate, dimethyl l-malate, l---malic acid dimethyl ester, 1,4-dimethyl 2s-2-hydroxybutanedioate, butanedioic acid, hydroxy-, dimethyl ester, 2s, dimethyl l---malate, pubchem6778, di-methyl s-malate, dimethyl s--malate
PubChem CID 10285815
IUPAC Name 1,4-dimethyl 2-hydroxybutanedioate
SMILES COC(=O)CC(O)C(=O)OC

Specifications

Color Yellow
Boiling Point 108°C
Quantity 5 g
Formula Weight 162.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl L-(-)-Malate