missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl L-Glutamate Hydrochloride 98.0+%, TCI America™
Supplier: TCI America D335325G
Specifications
| Dimethyl L-Glutamate Hydrochloride | |
| 93°C | |
| C7H13ClNO4 | |
| 25 g | |
| BPHCSYSXOKTCMA-UHFFFAOYNA-N | |
| 1,5-dimethyl 2-aminopentanedioate hydrochloridyl | |
| 12917568 | |
| ≥98.0% (N,T) |
| 23150-65-4 | |
| White | |
| MFCD00038879 | |
| l-glutamic acid dimethyl ester hydrochloride, h-glu ome-ome.hcl, dimethyl l-glutamate hydrochloride, s-dimethyl 2-aminopentanedioate hydrochloride, unii-185vc5x3pf, 1,5-dimethyl 2s-2-aminopentanedioate hydrochloride, h-glu ome-ome inverted exclamation mark currencyhcl, s-dimethyl 2-aminopentanedioate hcl, glutamic acid dimethyl ester hydrochloride, dimethyl glutamic acid hydrochloride | |
| [Cl].COC(=O)CCC(N)C(=O)OC | |
| 210.63 | |
| 211.64 | |
| Crystalline Powder |
Chemical Identifiers
| 23150-65-4 | |
| 210.63 | |
| BPHCSYSXOKTCMA-UHFFFAOYNA-N | |
| 12917568 | |
| [Cl].COC(=O)CCC(N)C(=O)OC |
| C7H13ClNO4 | |
| MFCD00038879 | |
| l-glutamic acid dimethyl ester hydrochloride, h-glu ome-ome.hcl, dimethyl l-glutamate hydrochloride, s-dimethyl 2-aminopentanedioate hydrochloride, unii-185vc5x3pf, 1,5-dimethyl 2s-2-aminopentanedioate hydrochloride, h-glu ome-ome inverted exclamation mark currencyhcl, s-dimethyl 2-aminopentanedioate hcl, glutamic acid dimethyl ester hydrochloride, dimethyl glutamic acid hydrochloride | |
| 1,5-dimethyl 2-aminopentanedioate hydrochloridyl |
Safety and Handling
TSCA : No