missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl isophthalate, 99%, Thermo Scientific™

Catalog No. p-118243
Change view
Click to view available options
Quantity:
25g
500g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC186510250 25g Plastic bottle
AC186515000 500g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC186510250 Supplier Thermo Scientific Chemicals Supplier No. 186510250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1459-93-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008433
InChI Key VNGOYPQMJFJDLV-UHFFFAOYSA-N
Synonym dimethyl isophthalate, dimethyl m-phthalate, isophthalic acid dimethyl ester, methyl isophthalate, 1,3-benzenedicarboxylic acid, dimethyl ester, morflex 1129, dimethyl 1,3-benzenedicarboxylate, dimethylisophthalate, methyl 3-carbomethoxy benzoate, isophthalic acid, dimethyl ester
PubChem CID 15088
IUPAC Name dimethyl benzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC=C1)C(=O)OC

Specifications

Boiling Point 124°C (16.0 hPa)
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H4(CO2CH3)2
Quantity 25g
Solubility Information Solubility in water: insoluble
Viscosity 2.14 mPa.s (99°C)
Formula Weight 194.19
Percent Purity 99%
Chemical Name or Material Dimethyl isophthalate, 99%
Show More Show Less
EINECSNumber 215-951-9
RTECSNumber NT2540000
TSCA TSCA

RUO – Research Use Only