missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Fumarate 98.0+%, TCI America™

Catalog No. F006925G
missing translation for 'changeView'
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F006925G 25 g
F0069500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. F006925G Supplier TCI America missing translation for 'supplierNo' F006925G

Chemical Identifiers

CAS 624-49-7
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00064438
InChI Key LDCRTTXIJACKKU-ONEGZZNKSA-N
Synonym dimethyl fumarate, tecfidera, e-dimethyl fumarate, fumaderm, dimethyl e-but-2-enedioate, fumaric acid, dimethyl ester, boletic acid dimethyl ester, fumaric acid dimethyl ester, methyl fumarate, dimethyl trans-ethylenedicarboxylate
PubChem CID 637568
ChEBI CHEBI:76004
IUPAC Name dimethyl (E)-but-2-enedioate
SMILES COC(=O)C=CC(=O)OC

Specifications

Melting Point 104°C
Color White
Boiling Point 205°C
Quantity 25 g
Formula Weight 144.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Fumarate
EINECSNumber (2)-1099&(2)-1142
RTECSNumber EM6125000
TSCA Yes