missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl DL-Glutamate Hydrochloride 98.0+%, TCI America™

Catalog No. D33055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33055G 5 g
D330525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33055G Supplier TCI America Supplier No. D33055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13515-99-6
Molecular Formula C7H14ClNO4
Molecular Weight (g/mol) 211.64
MDL Number MFCD00266895
InChI Key MFUPLHQOVIUESQ-UHFFFAOYNA-N
Synonym DL-Glutamic Acid Dimethyl Ester Hydrochloride, H-DL-Glu(OMe)-OMe.HCl
PubChem CID 3084392
IUPAC Name 1,5-dimethyl 2-aminopentanedioate hydrochloride
SMILES Cl.COC(=O)CCC(N)C(=O)OC

Specifications

Melting Point 154°C
Color White
Quantity 5 g
Formula Weight 211.64
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl DL-Glutamate Hydrochloride