missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Diselenide 98.0+%, TCI America™
$177.08 - $177.08
Chemical Identifiers
| CAS | 7101-31-7 |
|---|---|
| Molecular Formula | C2H6Se2 |
| Molecular Weight (g/mol) | 188.012 |
| InChI Key | VLXBWPOEOIIREY-UHFFFAOYSA-N |
| Synonym | dimethyl diselenide, dimethyldiselenide, methyl diselenide, diselenide, dimethyl, ch3se 2, methyldiselanyl methane, diselenobismethane, 1,2-dimethyldiselane, acmc-209ogy, methyl diselenide, 8ci |
| PubChem CID | 23496 |
| IUPAC Name | (methyldiselanyl)methane |
| SMILES | C[Se][Se]C |
Chemical Identifiers
| 7101-31-7 | |
| 188.012 | |
| dimethyl diselenide, dimethyldiselenide, methyl diselenide, diselenide, dimethyl, ch3se 2, methyldiselanyl methane, diselenobismethane, 1,2-dimethyldiselane, acmc-209ogy, methyl diselenide, 8ci | |
| (methyldiselanyl)methane |
| C2H6Se2 | |
| VLXBWPOEOIIREY-UHFFFAOYSA-N | |
| 23496 | |
| C[Se][Se]C |
Specifications
| 7101-31-7 | |
| 155°C | |
| 1 g | |
| dimethyl diselenide, dimethyldiselenide, methyl diselenide, diselenide, dimethyl, ch3se 2, methyldiselanyl methane, diselenobismethane, 1,2-dimethyldiselane, acmc-209ogy, methyl diselenide, 8ci | |
| C[Se][Se]C | |
| 188.012 | |
| 187.99 | |
| Liquid |
| -82°C | |
| C2H6Se2 | |
| 3440 | |
| VLXBWPOEOIIREY-UHFFFAOYSA-N | |
| (methyldiselanyl)methane | |
| 23496 | |
| ≥98.0% (GC) | |
| Dimethyl Diselenide |
Safety and Handling
TSCA : No