missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Dipropargylmalonate 98.0+%, TCI America™

Catalog No. D49635G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49635G 5 g
D49631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49635G Supplier TCI America Supplier No. D49635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63104-44-9
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
InChI Key AKISDSWMEPTCRL-UHFFFAOYSA-N
Synonym Dipropargylmalonic Acid Dimethyl Ester
PubChem CID 1546804
IUPAC Name dimethyl 2,2-bis(prop-2-ynyl)propanedioate
SMILES COC(=O)C(CC#C)(CC#C)C(=O)OC

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 5 g
Formula Weight 208.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Dipropargylmalonate