missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl D-Glutamate Hydrochloride 98.0+%, TCI America™
$84.70 - $246.43
Chemical Identifiers
| CAS | 27025-25-8 |
|---|---|
| Molecular Formula | C7H14ClNO4 |
| Molecular Weight (g/mol) | 211.642 |
| MDL Number | MFCD00235872 |
| InChI Key | MFUPLHQOVIUESQ-NUBCRITNSA-N |
| Synonym | dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride |
| PubChem CID | 12917567 |
| IUPAC Name | dimethyl (2R)-2-aminopentanedioate;hydrochloride |
| SMILES | COC(=O)CCC(C(=O)OC)N.Cl |
Chemical Identifiers
| 27025-25-8 | |
| 211.642 | |
| MFUPLHQOVIUESQ-NUBCRITNSA-N | |
| 12917567 | |
| COC(=O)CCC(C(=O)OC)N.Cl |
| C7H14ClNO4 | |
| MFCD00235872 | |
| dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride | |
| dimethyl (2R)-2-aminopentanedioate;hydrochloride |
Specifications
| 27025-25-8 | |
| White-Yellow | |
| MFCD00235872 | |
| dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride | |
| COC(=O)CCC(C(=O)OC)N.Cl | |
| 211.642 | |
| 211.64 | |
| Crystalline Powder |
| 91°C | |
| C7H14ClNO4 | |
| 5 g | |
| MFUPLHQOVIUESQ-NUBCRITNSA-N | |
| dimethyl (2R)-2-aminopentanedioate;hydrochloride | |
| 12917567 | |
| ≥98.0% (N,T) | |
| Dimethyl D-Glutamate Hydrochloride |
Safety and Handling
TSCA : No