missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl D-Glutamate Hydrochloride 98.0+%, TCI America™

Catalog No. D35605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35605G 5 g
D356025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35605G Supplier TCI America Supplier No. D35605G

Chemical Identifiers

CAS 27025-25-8
Molecular Formula C7H14ClNO4
Molecular Weight (g/mol) 211.642
MDL Number MFCD00235872
InChI Key MFUPLHQOVIUESQ-NUBCRITNSA-N
Synonym dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride
PubChem CID 12917567
IUPAC Name dimethyl (2R)-2-aminopentanedioate;hydrochloride
SMILES COC(=O)CCC(C(=O)OC)N.Cl

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 5 g
Formula Weight 211.64
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl D-Glutamate Hydrochloride