missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cyclohexylmalonate, 95%

Catalog No. AAH2628906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2628906 5 g
AAH2628903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2628906 Supplier Thermo Scientific Chemicals Supplier No. H2628906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 49769-76-8 | C11H18O4 | 214.261 g/mol

Used in developing a one-pot, two-step procedure for decarbalkoxylation of malonic esters. The effect of 15C5, other cations, and good nucleophiles was also studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in developing a one-pot, two-step procedure for decarbalkoxylation of malonic esters. The effect of 15C5, other cations, and good nucleophiles was also studied.

Solubility
Water soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 49769-76-8
Molecular Formula C11H18O4
Molecular Weight (g/mol) 214.261
MDL Number MFCD05666206
InChI Key CULITIDIVVTWIV-UHFFFAOYSA-N
Synonym dimethyl 2-cyclohexylmalonate, dimethyl cyclohexylmalonate, acmc-1asaj, dimethyl cyclohexylpropanedioate, cyclohexylmalonic acid dimethyl ester, cyclohexyl-malonic acid dimethyl ester, 1,3-dimethyl 2-cyclohexylpropanedioate, 2-cyclohexylmalonic acid dimethyl ester, dimethyl 2-cyclohexyl malonate, propanedioic acid, cyclohexyl-, dimethyl ester
PubChem CID 2793740
IUPAC Name dimethyl 2-cyclohexylpropanedioate
SMILES COC(=O)C(C1CCCCC1)C(=O)OC

Specifications

Refractive Index 1.4565
Quantity 5 g
Solubility Information Water soluble.
Formula Weight 214.26
Percent Purity 95%
Chemical Name or Material Dimethyl cyclohexylmalonate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only