missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl-beta-cyclodextrin, methylated beta-cyclodextrins, 96%

Catalog No. p-3757768
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC235530010 1 g
AC235530050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC235530010 Supplier Thermo Scientific Chemicals Supplier No. 235530010

CAS: 51166-71-3 | C56H98O35 | 1331.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 51166-71-3
Molecular Formula C56H98O35
Molecular Weight (g/mol) 1331.4
MDL Number MFCD00011616
InChI Key QGKBSGBYSPTPKJ-UZMKXNTCSA-N
Synonym 2,6-di-o-methyl-beta-cyclodextrin, unii-5fjp73e35o, dimethyl-beta-cyclodextrin, heptakis 2,6-o-dimethyl beta-cyclodextrin, heptakis-2,6-di-o-methyl-beta-cyclodextrin, heptakis 2,6-di-o-methyl-beta-cyclodextrin, dm-beta-cyd, dimethyl, a-cyclodextrin, 2,6-dme-beta-cyclodextrin, 2,6-di-o-methyl-beta-cd
PubChem CID 10171019
SMILES COCC1C2C(C(C(O1)OC3C(OC(C(C3O)OC)OC4C(OC(C(C4O)OC)OC5C(OC(C(C5O)OC)OC6C(OC(C(C6O)OC)OC7C(OC(C(C7O)OC)OC8C(OC(O2)C(C8O)OC)COC)COC)COC)COC)COC)COC)OC)O

Specifications

Melting Point 298.0°C to 310.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range (on dry basis) calculated
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition +163° (20°C c=1,water)
Specific Rotation + 163.00
Solubility Information Solubility in water: >330g/l (25°C). Other solubilities: <10g/l boiling water,>250g/l methanol,chloroform,dmso,dmf and,methylene chloride (25°C),<50g/l acetone (25°C)
Formula Weight 1331.4
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl-β-cyclodextrin, methylated β-cyclodextrins
Show More Show Less

RUO – Research Use Only