missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl chlorophosphate, 96%

Catalog No. AC426140050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426140050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426140050 Supplier Thermo Scientific Chemicals Supplier No. 426140050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 813-77-4 | C2H6ClO3P | 144.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 813-77-4
Molecular Formula C2H6ClO3P
Molecular Weight (g/mol) 144.49
MDL Number MFCD00117904
InChI Key NGFFLHMFSINFGB-UHFFFAOYSA-N
Synonym dimethyl phosphorochloridate, dimethyl chlorophosphate, phosphorochloridic acid, dimethyl ester, methyl phosphorochloridate, dimethylchlorophosphate, dimethyl chlorophosphonate, chlorodimethoxyphosphine oxide, chlorodimethylphosphate, dimethyl_chlorophosphate, pubchem19321
PubChem CID 101863
SMILES COP(Cl)(=O)OC

Specifications

Density 1.3400g/mL
Boiling Point 80°C (25.0 mmHg)
Flash Point >110°C
Assay Percent Range 95% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3O)2POCl
Quantity 5 g
Specific Gravity 1.34
IUPAC Name dimethyl phosphorochloridate
Formula Weight 144.49
Percent Purity 96%
Chemical Name or Material Dimethyl chlorophosphate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if swallowed.
Causes severe skin burns and eye damage.
Fatal in contact with skin.
Fatal if inhaled.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only