missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Chloromalonate 98.0+%, TCI America™

Catalog No. p-7128779
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54675G 5 g
D546725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54675G Supplier TCI America Supplier No. D54675G

Chemical Identifiers

CAS 28868-76-0
Molecular Formula C5H7ClO4
Molecular Weight (g/mol) 166.56
MDL Number MFCD00042837
InChI Key LNBQBURECUEBKZ-UHFFFAOYSA-N
Synonym Chloromalonic Acid Dimethyl Ester
PubChem CID 120070
IUPAC Name 1,3-dimethyl 2-chloropropanedioate
SMILES COC(=O)C(Cl)C(=O)OC

Specifications

Color Yellow
Boiling Point 208°C
Quantity 5 g
UN Number 3265
Formula Weight 166.56
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Chloromalonate