missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Bromomalonate 95.0+%, TCI America™

Catalog No. p-7127974
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
D38875G 5 g
D388725G 25 g
2 options

Catalog No. D38875G

Supplier: TCI America D38875G

Only null left

Chemical Identifiers

CAS 868-26-8
Molecular Formula C5H7BrO4
Molecular Weight (g/mol) 211.011
MDL Number MFCD00025865
InChI Key NEMOJKROKMMQBQ-UHFFFAOYSA-N
Synonym dimethyl bromomalonate, dimethyl 2-bromomalonate, dimethylbromomalonate, propanedioic acid, bromo-, dimethyl ester, 2-bromo dimethyl malonate, 1,3-dimethyl 2-bromopropanedioate, dimethylbromomalo-nate, acmc-209qbk, dimethyl-2-bromomalonate, dimethyl 2-bromomalonate #
PubChem CID 70087
IUPAC Name dimethyl 2-bromopropanedioate
SMILES COC(=O)C(C(=O)OC)Br

Specifications

Color Yellow
Boiling Point 225°C
Quantity 5 g
UN Number 3265
Formula Weight 211.01
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Bromomalonate