missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Benzylmalonate 95.0+%, TCI America™

Catalog No. D34325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34325G 5 g
D343225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34325G Supplier TCI America Supplier No. D34325G
Only null left

Chemical Identifiers

CAS 49769-78-0
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD01445471
InChI Key ZMYJSOIMFGAQRQ-UHFFFAOYSA-N
Synonym Benzylmalonic Acid Dimethyl Ester
PubChem CID 10900856
IUPAC Name 1,3-dimethyl 2-benzylpropanedioate
SMILES COC(=O)C(CC1=CC=CC=C1)C(=O)OC

Specifications

Color Colorless
Boiling Point 285°C
Quantity 5 g
Formula Weight 222.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Benzylmalonate