missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl allylmalonate, 97%

Catalog No. AAL0237618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0237618 50 g
AAL0237609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0237618 Supplier Thermo Scientific Chemicals Supplier No. L0237618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40637-56-7 | C8H12O4 | 172.18 g/mol

Dimethyl allylmalonate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl allylmalonate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Notes
Protect from moisture, heat. Incomaptible with oxidizing agents. Store in cool place.

Chemical Identifiers

CAS 40637-56-7
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00014959
InChI Key VZNFVLWVVHHMBG-UHFFFAOYSA-N
Synonym dimethyl allylmalonate, dimethyl 2-allylmalonate, propanedioic acid, 2-propenyl-, dimethyl ester, 1,3-dimethyl 2-prop-2-en-1-yl propanedioate, dimethyl allyl-malonate, acmc-1bnbn, dimethyl 2-allylmalonate #, 2-allyl-propanedioic acid dimethyl ester, 2-prop-2-enylpropanedioic acid dimethyl ester
PubChem CID 574826
IUPAC Name dimethyl 2-prop-2-enylpropanedioate
SMILES COC(=O)C(CC=C)C(=O)OC

Specifications

Melting Point <-80°C
Density 1.074
Boiling Point 207°C
Flash Point 95°C (203°F)
Refractive Index 1.435
Quantity 50 g
Beilstein 1772822
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material Dimethyl allylmalonate
EINECSNumber 255-017-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only