missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl adipimidate dihydrochloride, 97%

Catalog No. p-7045383
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1051504 2 g
AAL1051509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1051504 Supplier Thermo Scientific Chemicals Supplier No. L1051504
Only null left

CAS: 14620-72-5 | C8H18Cl2N2O2 | 245.144 g/mol

Dimethyl adipimidate dihydrochloride was used as a cross-linking reagent in the study of rhodopsin in sheep photoreceptor membrane

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl adipimidate dihydrochloride was used as a cross-linking reagent in the study of rhodopsin in sheep photoreceptor membrane

Solubility
Hydrolyses in water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, water.

Chemical Identifiers

CAS 14620-72-5
Molecular Formula C8H18Cl2N2O2
Molecular Weight (g/mol) 245.144
MDL Number MFCD00012573
InChI Key IEUUDEWWMRQUDS-UHFFFAOYSA-N
Synonym dimethyl adipimidate dihydrochloride, unii-who0f998gm, who0f998gm, adipimidic acid dimethyl ester dihydrochloride, dimethyl adipimidate hcl, dmai, acmc-209tig, bicl300, dimethyladipimidate dihydrochloride
PubChem CID 12876176
IUPAC Name dimethyl hexanediimidate;dihydrochloride
SMILES COC(=N)CCCCC(=N)OC.Cl.Cl

Specifications

Melting Point ∽215°C (decomposition)
Quantity 2 g
Sensitivity Moisture sensitive
Solubility Information Hydrolyses in water.
Formula Weight 245.15
Percent Purity 97%
Chemical Name or Material Dimethyl adipimidate dihydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-660-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only