missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl acetylmethylphosphonate, 97%

Catalog No. p-7043385
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0053706 5 g
AAL0053714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0053706 Supplier Thermo Scientific Chemicals Supplier No. L0053706
Only null left

CAS: 4202-14-6 | C5H11O4P | 166.113 g/mol

Dimethyl acetylmethylphosphonate is a Reagent used together with tosyl azide in a one-pot alkynylation of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl acetylmethylphosphonate is a Reagent used together with tosyl azide in a one-pot alkynylation of aldehydes.

Solubility
Difficult to mix.

Notes
Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 4202-14-6
Molecular Formula C5H11O4P
Molecular Weight (g/mol) 166.113
MDL Number MFCD00008769
InChI Key UOWIYNWMROWVDG-UHFFFAOYSA-N
Synonym dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone
PubChem CID 77872
IUPAC Name 1-dimethoxyphosphorylpropan-2-one
SMILES CC(=O)CP(=O)(OC)OC

Specifications

Density 1.202
Boiling Point 121°C to 123°C (11 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.439
Quantity 5 g
Beilstein 1766218
Solubility Information Difficult to mix.
Formula Weight 166.12
Percent Purity 97%
Chemical Name or Material Dimethyl acetylmethylphosphonate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 224-110-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only