missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Acetamide, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%, Honeywell Burdick & Jackson

Catalog No. p-7111735
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001857 6 x 1 L Glass Bottle
6001858 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001857 Supplier Honeywell Chemicals Supplier No. 0711L
Only null left

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Absorbance 0.3 AU max. at 275 nm (UV), 0.08 AU max. at 300 nm (UV), 0.005 AU max. at 350 nm (UV), 0.005 AU max. at 400 nm (UV)
CAS Max % 100.0000%
Quantity 6 x 1 L
Packaging Glass Bottle
Water 0.03% max.
Beilstein 1737614
Solubility Information Miscible in Water at 20°C
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material Dimethyl Acetamide, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%
ShelfLife 547 days from date of manufacture