missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Acetal 98.0+%, TCI America™

Catalog No. p-7126334
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D063325ML 25 mL
D0633500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D063325ML Supplier TCI America Supplier No. D063325ML
Only null left

Chemical Identifiers

CAS 534-15-6
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00008493
InChI Key SPEUIVXLLWOEMJ-UHFFFAOYSA-N
Synonym dimethyl acetal, acetaldehyde dimethyl acetal, ethane, 1,1-dimethoxy, dimethylacetal, dimethyl aldehyde, ethylidene dimethyl ether, methyl formyl, acetaldehyde, dimethyl acetal, acetaldehyde methyl acetal, 3-methyl-2,4-dioxapentane
PubChem CID 10795
IUPAC Name 1,1-dimethoxyethane
SMILES CC(OC)OC

Specifications

Melting Point -113°C
Color Colorless
Boiling Point 65°C
Quantity 25 mL
UN Number 2377
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Acetal
EINECSNumber (2)-0496&(2)-0568&(2)-0598
RTECSNumber AB2825000
TSCA Yes