missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 3,4-dihydroxypyrrole-2,5-dicarboxylate, 97%
CAS: 1632-19-5 | C8H9NO6 | 215.161 g/mol
$54.56 - $181.82
Chemical Identifiers
| CAS | 1632-19-5 |
|---|---|
| Molecular Formula | C8H9NO6 |
| Molecular Weight (g/mol) | 215.161 |
| MDL Number | MFCD00276587 |
| InChI Key | QCPBGHYMHYWRHG-UHFFFAOYSA-N |
| Synonym | dimethyl 3,4-dihydroxypyrrole-2,5-dicarboxylate, maybridge4_003392, acmc-20al76, 2,5-bis hydroxy methoxy methylidene pyrrolidine-3,4-dione, 2,5-dimethyl 3,4-dihydroxy-1h-pyrrole-2,5-dicarboxylate, methyl 3,4-dihydroxy-5-methoxycarbonyl pyrrole-2-carboxylate, 2z,5z-2,5-bis hydroxy methoxy methylidene pyrrolidine-3,4-dione, 1h-pyrrole-2,5-dicarboxylicacid, 3,4-dihydroxy-, 2,5-dimethyl ester |
| PubChem CID | 2727733 |
| IUPAC Name | dimethyl 3,4-dihydroxy-1H-pyrrole-2,5-dicarboxylate |
| SMILES | COC(=O)C1=C(C(=C(N1)C(=O)OC)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1923803
|
Thermo Scientific Chemicals
A1923803 |
1 g |
Each for $54.56
|
|
|||||
|
AAA1923806
|
Thermo Scientific Chemicals
A1923806 |
5 g |
Each for $181.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1632-19-5 | |
| 215.161 | |
| QCPBGHYMHYWRHG-UHFFFAOYSA-N | |
| 2727733 | |
| COC(=O)C1=C(C(=C(N1)C(=O)OC)O)O |
| C8H9NO6 | |
| MFCD00276587 | |
| dimethyl 3,4-dihydroxypyrrole-2,5-dicarboxylate, maybridge4_003392, acmc-20al76, 2,5-bis hydroxy methoxy methylidene pyrrolidine-3,4-dione, 2,5-dimethyl 3,4-dihydroxy-1h-pyrrole-2,5-dicarboxylate, methyl 3,4-dihydroxy-5-methoxycarbonyl pyrrole-2-carboxylate, 2z,5z-2,5-bis hydroxy methoxy methylidene pyrrolidine-3,4-dione, 1h-pyrrole-2,5-dicarboxylicacid, 3,4-dihydroxy-, 2,5-dimethyl ester | |
| dimethyl 3,4-dihydroxy-1H-pyrrole-2,5-dicarboxylate |
Specifications
| 1632-19-5 | |
| 1.248 | |
| >110°C (230°F) | |
| MFCD00276587 | |
| dimethyl 3,4-dihydroxypyrrole-2,5-dicarboxylate, maybridge4_003392, acmc-20al76, 2,5-bis hydroxy methoxy methylidene pyrrolidine-3,4-dione, 2,5-dimethyl 3,4-dihydroxy-1h-pyrrole-2,5-dicarboxylate, methyl 3,4-dihydroxy-5-methoxycarbonyl pyrrole-2-carboxylate, 2z,5z-2,5-bis hydroxy methoxy methylidene pyrrolidine-3,4-dione, 1h-pyrrole-2,5-dicarboxylicacid, 3,4-dihydroxy-, 2,5-dimethyl ester | |
| COC(=O)C1=C(C(=C(N1)C(=O)OC)O)O | |
| 215.161 | |
| 215.16 | |
| Dimethyl 3,4-dihydroxypyrrole-2,5-dicarboxylate |
| 209°C to 212°C | |
| 268°C to 272°C | |
| C8H9NO6 | |
| 1 g | |
| QCPBGHYMHYWRHG-UHFFFAOYSA-N | |
| dimethyl 3,4-dihydroxy-1H-pyrrole-2,5-dicarboxylate | |
| 2727733 | |
| 97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only